Assay Details
| Gene Symbol: LOC4333930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774854.1 | XP_015630340.1 | 1 - 2 | 234 | 124 | |
| GenBank mRNA | AB109891.1 | - | 1 - 2 | 88 | 124 | |
| AK072482.1 | - | 1 - 2 | 136 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015774854.1 transcripts.
Set Membership: |
|

LOC4333930
Genomic Map