Assay Details
| Gene Symbol: LOC4334101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777569.2 | XP_015633055.1 | 8 - 9 | 959 | 87 | |
| XM_015777570.2 | XP_015633056.1 | 8 - 9 | 900 | 87 | ||
| GenBank mRNA | AF073697.1 | - | 8 - 9 | 876 | 87 | |
| AK071279.1 | - | 8 - 9 | 851 | 87 | ||
More Information
Set Membership: |
|

LOC4334101
Genomic Map