Assay Details
| Gene Symbol: LOC4334171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775934.2 | XP_015631420.1 | 14 - 15 | 2919 | 72 | |
| GenBank mRNA | AK063766.1 | - | 13 - 14 | 2570 | 72 | |
| KT199368.1 | - | 9 - 10 | 1686 | 72 | ||
More Information
Set Membership: |
|

LOC4334171
Genomic Map