Assay Details
| Gene Symbol: LOC4334171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775934.2 | XP_015631420.1 | 6 - 7 | 1361 | 60 | |
| GenBank mRNA | AK063766.1 | - | 5 - 6 | 1012 | 60 | |
| KT199368.1 | - | 1 - 2 | 128 | 60 | ||
More Information
Set Membership: |
|

LOC4334171
Genomic Map