Assay Details
| Gene Symbol: LOC4337272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779836.2 | XP_015635322.1 | 10 - 11 | 1111 | 56 | |
| GenBank mRNA | AK063706.1 | - | 10 - 11 | 969 | 56 | |
| AK065491.1 | - | 10 - 11 | 969 | 56 | ||
| AK099252.1 | - | 10 - 11 | 941 | 56 | ||
More Information
Set Membership: |
|

LOC4337272
Genomic Map