Assay Details
| Gene Symbol: LOC4337272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779836.2 | XP_015635322.1 | 3 - 4 | 501 | 97 | |
| GenBank mRNA | AK063706.1 | - | 3 - 4 | 359 | 97 | |
| AK065491.1 | - | 3 - 4 | 360 | 97 | ||
More Information
Set Membership: |
|

LOC4337272
Genomic Map