Assay Details
| Gene Symbol: LOC4338051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782226.2 | XP_015637712.1 | 2 - 3 | 962 | 69 | |
| GenBank mRNA | AB126090.1 | - | 2 - 3 | 697 | 69 | |
| AK071272.1 | - | 2 - 3 | 928 | 69 | ||
| AY275180.1 | - | 2 - 3 | 703 | 69 | ||
More Information
Set Membership: |
|

LOC4338051
Genomic Map