Assay Details
Gene Symbol: LOC4338126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785207.2 | XP_015640693.1 | 1 - 2 | 629 | 78 | |
XM_015785209.2 | XP_015640695.1 | 2 - 3 | 536 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
