Assay Details
Gene Symbol: LOC4338126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785207.2 | XP_015640693.1 | 4 - 5 | 1285 | 95 | |
XM_015785209.2 | XP_015640695.1 | 5 - 6 | 1192 | 95 | ||
GenBank mRNA | AK122045.1 | - | 5 - 6 | 1094 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
