Assay Details
Gene Symbol: LOC4350997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK288106.1 | - | 3173 | 80 | ||
AK318564.1 | - | 3173 | 80 |
Gene Symbol: LOC4327027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK242245.1 | - | 1686 | 80 |
Gene Symbol: LOC4331124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK242265.1 | - | 2063 | 80 | ||
AK288092.1 | - | 2064 | 80 |
Gene Symbol: LOC4339877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK318560.1 | - | 2597 | 80 |
Gene Symbol: LOC4344664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK242017.1 | - | 1431 | 80 |
