Assay Details
Gene Symbol: LOC4339021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785253.2 | XP_015640739.1 | 1 - 2 | 194 | 148 | |
GenBank mRNA | AK111917.1 | - | 1 - 2 | 165 | 148 | |
AK111946.1 | - | 1 - 2 | 144 | 148 | ||
AK111972.1 | - | 1 - 2 | 87 | 148 | ||
AK112021.1 | - | 1 - 2 | 81 | 148 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015785253.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
