Assay Details
Gene Symbol: LOC4339021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785253.2 | XP_015640739.1 | 3 - 4 | 429 | 77 | |
XM_015785254.2 | XP_015640740.1 | 3 - 4 | 681 | 77 | ||
GenBank mRNA | AK111917.1 | - | 3 - 4 | 400 | 77 | |
AK111946.1 | - | 3 - 4 | 379 | 77 | ||
AK111972.1 | - | 3 - 4 | 322 | 77 | ||
AK112021.1 | - | 3 - 4 | 316 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
