Assay Details
Gene Symbol: LOC4339021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785253.2 | XP_015640739.1 | 6 - 7 | 687 | 111 | |
XM_015785254.2 | XP_015640740.1 | 6 - 7 | 939 | 111 | ||
GenBank mRNA | AK111917.1 | - | 6 - 7 | 658 | 111 | |
AK111946.1 | - | 6 - 7 | 637 | 111 | ||
AK111972.1 | - | 6 - 7 | 580 | 111 | ||
AK112021.1 | - | 6 - 7 | 574 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
