Assay Details
Gene Symbol: LOC4339021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785253.2 | XP_015640739.1 | 7 - 8 | 756 | 114 | |
XM_015785254.2 | XP_015640740.1 | 7 - 8 | 1008 | 114 | ||
GenBank mRNA | AK111917.1 | - | 7 - 8 | 727 | 114 | |
AK111946.1 | - | 7 - 8 | 706 | 114 | ||
AK111972.1 | - | 7 - 8 | 649 | 114 | ||
AK112021.1 | - | 7 - 8 | 643 | 114 | ||
BT130297.1 | - | 1 - 2 | 70 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
