Assay Details
Gene Symbol: LOC4339021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785253.2 | XP_015640739.1 | 8 - 9 | 843 | 91 | |
XM_015785254.2 | XP_015640740.1 | 8 - 9 | 1095 | 91 | ||
GenBank mRNA | AK111917.1 | - | 8 - 9 | 814 | 91 | |
AK111946.1 | - | 8 - 9 | 793 | 91 | ||
AK111972.1 | - | 8 - 9 | 736 | 91 | ||
AK112021.1 | - | 8 - 9 | 730 | 91 | ||
BT130297.1 | - | 2 - 3 | 157 | 91 | ||
BT132603.1 | - | 2 - 3 | 145 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
