Assay Details
Gene Symbol: LOC4339026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784239.2 | XP_015639725.1 | 3 - 4 | 2917 | 104 | |
XM_015784240.2 | XP_015639726.1 | 4 - 5 | 2617 | 104 | ||
GenBank mRNA | AK099608.1 | - | 3 - 4 | 2865 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
