Assay Details
| Gene Symbol: LOC4339429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783525.2 | XP_015639011.1 | 11 - 12 | 1164 | 138 | |
| XM_015783526.2 | XP_015639012.1 | 12 - 13 | 1161 | 138 | ||
| GenBank mRNA | AK106477.1 | - | 12 - 13 | 981 | 138 | |
More Information
Set Membership: |
|

LOC4339429
Genomic Map