Assay Details
| Gene Symbol: LOC4339429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783525.2 | XP_015639011.1 | 4 - 5 | 610 | 84 | |
| XM_015783526.2 | XP_015639012.1 | 5 - 6 | 607 | 84 | ||
| GenBank mRNA | AK106477.1 | - | 5 - 6 | 427 | 84 | |
More Information
Set Membership: |
|

LOC4339429
Genomic Map