Assay Details
| Gene Symbol: LOC4340102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788795.2 | XP_015644281.1 | 8 - 9 | 1688 | 147 | |
| GenBank mRNA | AK103630.1 | - | 8 - 9 | 1664 | 147 | |
More Information
Set Membership: |
|

LOC4340102
Genomic Map