Assay Details
| Gene Symbol: LOC4340221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785722.2 | XP_015641208.2 | 11 - 12 | 1126 | 85 | |
| GenBank mRNA | AK068888.1 | - | 11 - 12 | 952 | 85 | |
| BT132410.1 | - | 1 - 2 | 95 | 85 | ||
More Information
Set Membership: |
|

LOC4340221
Genomic Map