Assay Details
Gene Symbol: LOC4340242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788141.2 | XP_015643627.1 | 1 - 2 | 424 | 149 | |
XM_015788143.2 | XP_015643629.1 | 1 - 2 | 424 | 149 | ||
GenBank mRNA | AK066174.1 | - | 1 - 2 | 308 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788141.2, XM_015788143.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
