Assay Details
Gene Symbol: LOC4340242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788141.2 | XP_015643627.1 | 2 - 3 | 2551 | 114 | |
XM_015788143.2 | XP_015643629.1 | 2 - 3 | 2551 | 114 | ||
GenBank mRNA | AK066174.1 | - | 2 - 3 | 2435 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
