Assay Details
Gene Symbol: LOC4340242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788143.2 | XP_015643629.1 | 4 - 5 | 2771 | 66 | |
GenBank mRNA | AK066174.1 | - | 4 - 5 | 2655 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
