Assay Details
Gene Symbol: LOC4340242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788141.2 | XP_015643627.1 | 5 - 6 | 2839 | 94 | |
XM_015788143.2 | XP_015643629.1 | 5 - 6 | 2836 | 94 | ||
GenBank mRNA | AK066174.1 | - | 5 - 6 | 2720 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
