Assay Details
| Gene Symbol: LOC4340366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787957.2 | XP_015643443.1 | 3 - 4 | 1027 | 69 | |
| GenBank mRNA | AK073349.1 | - | 3 - 4 | 1022 | 69 | |
More Information
Set Membership: |
|

LOC4340366
Genomic Map