Assay Details
| Gene Symbol: LOC4341189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787602.2 | XP_015643088.1 | 7 - 8 | 1681 | 80 | |
| XM_015787603.1 | XP_015643089.1 | 6 - 7 | 1586 | 80 | ||
| GenBank mRNA | AK099993.1 | - | 7 - 8 | 1646 | 80 | |
More Information
Set Membership: |
|

LOC4341189
Genomic Map