Assay Details
| Gene Symbol: LOC4337621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783967.2 | XP_015639453.1 | 1 - 2 | 468 | 76 | |
| GenBank mRNA | AK058320.1 | - | 1 - 2 | 293 | 76 | |
| AK121775.1 | - | 1 - 2 | 445 | 76 | ||
More Information
Set Membership: |
|

LOC4337621
Genomic Map