Assay Details
| Gene Symbol: LOC4352284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK242914.1 | - | 3045 | 85 | ||
| Gene Symbol: LOC4338485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784467.2 | XP_015639953.1 | 1 - 2 | 130 | 85 | |
| GenBank mRNA | AF529175.1 | - | 1 - 2 | 93 | 85 | |
| AK061627.1 | - | 1 - 2 | 104 | 85 | ||
| AK103081.1 | - | 1 - 2 | 104 | 85 | ||
| AK104191.1 | - | 1 - 2 | 104 | 85 | ||
| BT129368.1 | - | 1 - 2 | 99 | 85 | ||
| BT132274.1 | - | 1 - 2 | 97 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784467.2 transcripts.
Set Membership: |
|

LOC4352284
Genomic Map