Assay Details
| Gene Symbol: LOC4338485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784467.2 | XP_015639953.1 | 3 - 4 | 284 | 102 | |
| GenBank mRNA | AF529175.1 | - | 3 - 4 | 247 | 102 | |
| AK061627.1 | - | 3 - 4 | 258 | 102 | ||
| AK103081.1 | - | 3 - 4 | 258 | 102 | ||
| AK104191.1 | - | 3 - 4 | 258 | 102 | ||
| BT132274.1 | - | 3 - 4 | 251 | 102 | ||
| Gene Symbol: LOC4352284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK242914.1 | - | 2891 | 102 | ||
More Information
Set Membership: |
|

LOC4338485
Genomic Map