Assay Details
| Gene Symbol: LOC4338545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783813.2 | XP_015639299.1 | 5 - 6 | 826 | 81 | |
| GenBank mRNA | AK073358.1 | - | 5 - 6 | 793 | 81 | |
| AK073379.1 | - | 5 - 6 | 793 | 81 | ||
More Information
Set Membership: |
|

LOC4338545
Genomic Map