Assay Details
| Gene Symbol: LOC4339546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784533.2 | XP_015640019.1 | 3 - 4 | 737 | 67 | |
| GenBank mRNA | AK061681.1 | - | 3 - 4 | 678 | 67 | |
| AK071143.1 | - | 3 - 4 | 737 | 67 | ||
| KF214533.1 | - | 3 - 4 | 666 | 67 | ||
More Information
Set Membership: |
|

LOC4339546
Genomic Map