Assay Details
Gene Symbol: LOC4336044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK241494.1 | - | 1818 | 77 | ||
AK287771.1 | - | 1819 | 77 |
Gene Symbol: LOC4340693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787247.2 | XP_015642733.1 | 1 | 157 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
