Assay Details
| Gene Symbol: LOC4341457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786499.2 | XP_015641985.1 | 4 - 5 | 1127 | 84 | |
| GenBank mRNA | AK106474.1 | - | 4 - 5 | 689 | 84 | |
More Information
Set Membership: |
|

LOC4341457
Genomic Map