Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 11 - 12 | 2221 | 76 | |
| GenBank mRNA | AB096015.1 | - | 11 - 12 | 2084 | 76 | |
| AB120430.1 | - | 11 - 12 | 2227 | 76 | ||
| AK058530.1 | - | 4 - 5 | 589 | 76 | ||
| AK069977.1 | - | 12 - 13 | 1861 | 76 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map