Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 13 - 14 | 2460 | 133 | |
| GenBank mRNA | AB096015.1 | - | 13 - 14 | 2323 | 133 | |
| AB120430.1 | - | 13 - 14 | 2466 | 133 | ||
| AK058530.1 | - | 6 - 7 | 828 | 133 | ||
| AK069977.1 | - | 14 - 15 | 2100 | 133 | ||
| U34283.1 | - | 1 - 2 | 142 | 133 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map