Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 14 - 15 | 2536 | 69 | |
| GenBank mRNA | AB096015.1 | - | 14 - 15 | 2399 | 69 | |
| AB120430.1 | - | 14 - 15 | 2542 | 69 | ||
| AK058530.1 | - | 7 - 8 | 904 | 69 | ||
| AK069977.1 | - | 15 - 16 | 2176 | 69 | ||
| U34283.1 | - | 2 - 3 | 218 | 69 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map