Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 3 - 4 | 1187 | 95 | |
| GenBank mRNA | AB096015.1 | - | 3 - 4 | 1050 | 95 | |
| AB120430.1 | - | 3 - 4 | 1193 | 95 | ||
| AK069977.1 | - | 4 - 5 | 827 | 95 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map