Assay Details
| Gene Symbol: LOC4341695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788508.2 | XP_015643994.1 | 4 - 5 | 1258 | 81 | |
| GenBank mRNA | AB096015.1 | - | 4 - 5 | 1121 | 81 | |
| AB120430.1 | - | 4 - 5 | 1264 | 81 | ||
| AK069977.1 | - | 5 - 6 | 898 | 81 | ||
More Information
Set Membership: |
|

LOC4341695
Genomic Map