Assay Details
Gene Symbol: LOC4341859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788997.2 | XP_015644483.1 | 1 - 2 | 418 | 68 | |
XM_015788998.2 | XP_015644484.1 | 1 - 2 | 419 | 68 | ||
GenBank mRNA | AK058781.1 | - | 1 - 2 | 397 | 68 | |
AK110977.1 | - | 1 - 2 | 387 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788997.2, XM_015788998.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
