Assay Details
| Gene Symbol: LOC4341859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788997.2 | XP_015644483.1 | 4 - 5 | 2297 | 101 | |
| XM_015788998.2 | XP_015644484.1 | 4 - 5 | 691 | 101 | ||
| GenBank mRNA | AK110977.1 | - | 4 - 5 | 659 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788997.2, XM_015788998.2 transcripts.
Set Membership: |
|

LOC4341859
Genomic Map