Assay Details
Gene Symbol: LOC4341859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788997.2 | XP_015644483.1 | 5 - 6 | 2394 | 74 | |
XM_015788998.2 | XP_015644484.1 | 5 - 6 | 788 | 74 | ||
GenBank mRNA | AK110977.1 | - | 5 - 6 | 756 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
