Assay Details
| Gene Symbol: LOC4341859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788997.2 | XP_015644483.1 | 6 - 7 | 2476 | 63 | |
| XM_015788998.2 | XP_015644484.1 | 6 - 7 | 870 | 63 | ||
| GenBank mRNA | AK110977.1 | - | 6 - 7 | 838 | 63 | |
More Information
Set Membership: |
|

LOC4341859
Genomic Map