Assay Details
| Gene Symbol: LOC4341997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787519.1 | XP_015643005.1 | 12 - 13 | 1284 | 85 | |
| XM_015787521.1 | XP_015643007.1 | 11 - 12 | 1120 | 85 | ||
| GenBank mRNA | AK065784.1 | - | 12 - 13 | 1176 | 85 | |
| AK101836.1 | - | 12 - 13 | 1275 | 85 | ||
| AK119551.1 | - | 12 - 13 | 1168 | 85 | ||
More Information
Set Membership: |
|

LOC4341997
Genomic Map