Assay Details
| Gene Symbol: LOC4341997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787519.1 | XP_015643005.1 | 1 - 2 | 172 | 70 | |
| GenBank mRNA | AK101836.1 | - | 1 - 2 | 163 | 70 | |
| AK119551.1 | - | 1 - 2 | 56 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787519.1 transcripts.
Set Membership: |
|

LOC4341997
Genomic Map