Assay Details
Gene Symbol: LOC4342403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791887.2 | XP_015647373.1 | 29 - 30 | 4187 | 136 | |
XM_015791888.2 | XP_015647374.1 | 29 - 30 | 4121 | 136 | ||
GenBank mRNA | AK062246.1 | - | 1 - 2 | 48 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
