Assay Details
| Gene Symbol: LOC4342403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791887.2 | XP_015647373.1 | 33 - 34 | 4741 | 81 | |
| XM_015791888.2 | XP_015647374.1 | 33 - 34 | 4675 | 81 | ||
| GenBank mRNA | AK062246.1 | - | 5 - 6 | 602 | 81 | |
More Information
Set Membership: |
|

LOC4342403
Genomic Map