Assay Details
| Gene Symbol: LOC4342471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792394.2 | XP_015647880.1 | 3 - 4 | 1052 | 143 | |
| GenBank mRNA | AB583903.1 | - | 3 - 4 | 802 | 143 | |
| AK111609.1 | - | 3 - 4 | 1037 | 143 | ||
More Information
Set Membership: |
|

LOC4342471
Genomic Map