Assay Details
| Gene Symbol: LOC4342593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792086.1 | XP_015647572.1 | 2 - 3 | 313 | 76 | |
| GenBank mRNA | AK059196.1 | - | 2 - 3 | 304 | 76 | |
| AK121423.1 | - | 2 - 3 | 305 | 76 | ||
| BT129316.1 | - | 2 - 3 | 316 | 76 | ||
| BT129433.1 | - | 2 - 3 | 301 | 76 | ||
| D21836.1 | - | 2 - 3 | 267 | 76 | ||
More Information
Set Membership: |
|

LOC4342593
Genomic Map