Assay Details
| Gene Symbol: LOC4343045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789605.2 | XP_015645091.1 | 17 - 18 | 2225 | 95 | |
| XM_015789606.2 | XP_015645092.2 | 17 - 18 | 2275 | 95 | ||
| GenBank mRNA | AK121014.1 | - | 17 - 18 | 2232 | 95 | |
More Information
Set Membership: |
|

LOC4343045
Genomic Map