Assay Details
Gene Symbol: LOC4343109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790716.2 | XP_015646202.1 | 1 - 2 | 254 | 128 | |
GenBank mRNA | AK099317.1 | - | 1 - 2 | 207 | 128 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015790716.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
